Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carbol fuchsin (Kinyoun’s) stain for bacteriology, for AFB (acid fast bacteria)

Supplier: HARDY DIAGNOSTICS

CF008
76045-454EA 26.86 USD InStock
76045-454
Carbol fuchsin (Kinyoun’s) stain for bacteriology, for AFB (acid fast bacteria)
Carbol fuchsin
Carbol Fuchsin, Kinyoun for AFB stain, for Mycobacteria

An aqueous solution of basic fuchsin, phenol, and reagent alcohol for the “Cold Stain” method.


Formula: C₂₁H₂₂ClN₃
MW: 351.88 g/mol
MDL Number: MFCD00013129
CAS Number: 4197-24-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty