Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Urea 99.0-100.5% (by titrimetric analysis), prilled USP

Supplier: CHEM IMPEX INTL INC MS
Synonyms: Carbonyl diamideCarbamideCarbonyldiamideDiaminomethanone

00309-5KG 00309-1KG
102572-736EA 37.57 USD InStock
102572-736 102587-456
Urea 99.0-100.5% (by titrimetric analysis), prilled USP
Urea


Formula: NH₂CONH₂
MW: 60.06 g/mol
Melting Pt: 132…135 °C
MDL Number: MFCD00008022
CAS Number: 57-13-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/70/46/2297046.jpg
Order Now
Specification Test Results
Appearance White powder or pastilles
Melting Point 132-135 °C
Purity Limit 99.0-100.5% (Titration)
Grade USP
Storage Temperature Room Temperature

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty