Would you like to log in again?
Muscle Histology Bundle
Supplier: Avantor
470230-958EA 635.09 USD InStock
470230-958
Muscle Histology Bundle
Models Human Biology Models
Compare Muscular Structure With Models And Slides
  • Buy as a set and save
The Ward's Muscle types models represent small sections of muscle fiber that are designed based on structure viewed from microscope slides. Each of the three models represents a different muscle type (Cardiac, smooth, and skeletal) and show features common to all and unique to each. an included key helps identify specific structures and allowing for easy comparison between the types. The three included microscope slides are Ward's own high quality preparations. The first slide allows for comparison of each muscle type is section. Students can compare structures seen on the slide with the model for that muscle type. A second slide shows skeleton muscle with the fibers teased apart and stained to show striations and the nuclei that make this type of muscle so distinct. The final slide shows motor neurons, the cells that signal muscular action.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty