Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 10% in aqueous solution

Supplier: Electron Microscopy Sciences
Synonyms: Muriatic acid

16765
101410-690EA 57.14 USD InStock
101410-690
Hydrochloric acid 10% in aqueous solution
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/35/28984835.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty