Would you like to log in again?
Biohazard Authorized Personnel Only Sign, Brady Worldwide
Supplier: Brady
62812
76046-534EA 49.78 USD InStock
76046-534
Biohazard Authorized Personnel Only Sign, Brady Worldwide
Signs
Chemical, Biohazard, and Hazardous Material signs.
  • Graphics are permanently embedded within the sign panel, allowing for use in the most severe environments
  • Excellent chemical and abrasion resistance
  • Ideal for both indoor and outdoor applications
  • Service temperature of –40 to 90 °C (–40 to 193 °F)
Extremely durable B-120 Premium Fiberglass will not chip, fade, rust, shatter, or peel for up to 25 years.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty