Would you like to log in again?
BRIGHTSORB™ Chemical Absorbent Socks, Brady Worldwide
Supplier: Brady
148626 148628
76046-565CS 203.67 USD InStock
76046-565 76046-570
BRIGHTSORB™ Chemical Absorbent Socks, Brady Worldwide
Spill Control Materials
BRIGHTSORB™ High Visibility absorbents feature bright safety yellow coloring for easy separation of hazardous waste
  • Moldable SOCs wrap around machinery and conform to uneven surfaces to prevent leaks from reaching walkways
  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • BRIGHTSORB™ High Visibility series combine safety signage and absorbency in one product
  • Slip, trip, and fall hazard warning printed on the absorbent alerts employees to potential danger
Ideal in areas with a high risk of slips, trips, and falls.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty