Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
6.5 Gallon Specialty Spill Kits, Brady Worldwide
Supplier: Brady
148856 148857 148858
76000-808KT 369.02 USD InStock
76000-808 76000-810 76000-812
6.5 Gallon Specialty Spill Kits, Brady Worldwide
Spill Control Materials
Spill Kits combine absorbent pads, accessories, PPE, and instruction guidelines for reliable preparation in an emergency
  • Spill kit contains surfactant-treated absorbents and contents designed to pick up battery acid while remaining chemically inert
  • Reusable screw top bucket keeps contents clean and easily accessible
The Formaldehyde Spill Kit includes (10) 15" x 19" pads; (1) 1 lb. bag granular; (2) disposal bags; (1) 2 lb. acid neutralizer; (1) respirator; (1) goggles; (1) neorprene gloves; (1) scoop and scraper.
The Bodily Fluids Spill Kit includes (10) 15" x 19" barrier backed pads; (2) disposal bags; (1) 1 lb. bag granular; (1) set sanitary wipes; (1) neoprene apron; (1) scoop and scraper; (1) goggles; (1) pair neoprene gloves.
The Battery Acid Spill Kit includes (10) 15" x 19" pads; (2) 3" x 4' socks; (2) disposal bags; (1) 1 lb. bag granular; (1) neoprene apron; (1) pair neoprene gloves; (1) scoop and scraper; (1) face shield; (1) pair goggles.
Position them around the facility to ensure all of the necessary materials are within reach should a spill occur.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty