Would you like to log in again?
CleanLift® Repositionable Vinyl Cloth Labels, White, Brady
Supplier: Brady
CleanLift
62678 114227 62683 62686 114228 62681 62679 114223 114224 64391 62680 62690 62685 62689 62682 62688 114221 62687 62684 114225
76046-417RL 374.66 USD InStock
76046-417 76046-517 76046-528 76046-419 76046-519 76046-509 76046-530 76046-521 76046-423 76046-511 76046-533 76046-412 76046-523 76046-425 76046-513 76046-525 76046-415 76046-515 76046-537 76046-526
CleanLift® Repositionable Vinyl Cloth Labels, White, Brady
Labels
B-498 CleanLift® series vinyl cloth labels are available in open 1 or 3" cores.
  • The adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning
  • Sticks to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • Semi-gloss finish for crisp, clean look
Printable, repositionable vinyl cloth labels for thermal transfer printers.
Note: This material is UL recognized with its respective ribbon.
It is used for barcode labeling, cable and wire marking, circuit board ID - through hole top, component and equipment ID, general and industrial ID.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty