Would you like to log in again?
SP Wilmad-LabGlass Precision Glass Sleeve Stirrer Bearings, SP Industries
Supplier: SP Industries
LG-9411-142
14203-462EA 204.71 USD InStock
14203-462
SP Wilmad-LabGlass Precision Glass Sleeve Stirrer Bearings, SP Industries
Stirrers Stirring Tools
These glass sleeve bearings have a 100 mm bore and are specially processed for precision fit with a ground shaft.
They feature a small grease cup at the top and a full-length inner joint at lower end. Use with stirrer shaft (LG-9500), glass stirrer blade (LG-9505), and/or PTFE blade (LG-9510T).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty