Would you like to log in again?
Rappaport Vassiliadis Broth CS Tubed Culture Media, Biomerieux
Supplier: bioMérieux
42073
95060-760CS 76.8 USD InStock
95060-760
Rappaport Vassiliadis Broth CS Tubed Culture Media, Biomerieux
Media for Microbiology Prepared Media for Microbiology
Rappaport Vassiliadis broth (containing malachite green and magnesium chloride) is a selective enrichment broth used for the detection of Salmonella in food
  • For detection of Salmonella in food
  • 20 x 10 mL tubes
Rappaport Vassiliadis (RV) broth contains a peptone base which favors the growth of Salmonella. Also contains potassium dihydrogen phosphate for improved pH stability, and magnesium chloride at a concentration which reduces the toxic effect of malachite green and ensures selectivity of the medium.
It is generally used in parallel with Selenite Cystine (SC) broth.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty