Would you like to log in again?
Tryptic Soy Agar Tubed Culture Media, Biomerieux
Supplier: bioMérieux
P1120 T8076-C T8076-C
95061-592CS 438.6 USD InStock
95061-592 95061-436 CA-95061-592
Tryptic Soy Agar Tubed Culture Media, Biomerieux
Media for Microbiology Prepared Media for Microbiology
Tryptic Soy Agar is a general-purpose solid medium used for the cultivation and enumeration of a wide variety of fastidious and nonfastidious microorganisms.
  • General purpose medium for the cultivation of a wide variety of microorganisms
  • Multiple sizes available
Upon receipt, store bottles and tubes according to the package label in an upright position and in the dark. Do not freeze. Allow warming of medium to room temperature before inoculation. Media can be inoculated up to the expiration date and incubated for the appropriate incubation period. Expiration date applies to the product in an unopened container stored as directed. See package label.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty