Would you like to log in again?
Neulog Distillation Kit
Supplier: Neulog
DSTKIT
470229-962KT 133.55 USD InStock
470229-962
Neulog Distillation Kit
Educational Classroom Kits and Activities
Hardware to do simple distillation experiments.
  • Investigate the distillation process with NeuLog sensors
  • Requires NeuLog temperature logger sensor
Distillation is a method for separating mixtures based on their different boiling points. The separation is conducted by boiling and condensation. What happens to the temperature of a liquid during the boiling process? You might think that the temperature goes up smoothly; but this is not so. The graph of temperature against time is called a heating curve. In general; the longer the heating continues; the temperature goes up. However; there are horizontal flat parts on the graph which happen when there is a change of state. The plateaus are also called phase changes.
With this kit you will be able to investigate this fascinating behavior through measurements of both pure water and a mixture of water and alcohol or acetone. This kit allows students to visualize heating curves and to understand the concept of distillation.
Delivery information: Still head, 25 ml round bottom flask, condenser, receiver, alcohol lamp, two 50 ml beakers, utility stand, right angle clamp, extension clamp, four boiling stones.
NeuLog sensor required (not included): temperature logger sensor.
Caution: For educational use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty