Would you like to log in again?
Neulog Gas Kit
Supplier: Neulog
GASKIT
470229-966KT 109.75 USD InStock
470229-966
Neulog Gas Kit
Educational Classroom Kits and Activities
Hardware to do NeuLog experiments about gas laws.
  • Allows for study of gas pressure as a function of volume, gas pressure as a function of temperature, linear fitting on a graph, and extrapolation
The Neulog Gas Kit provides all of the necessary tools to study gas pressure as a function of volume, gas pressure as a function of temperature, linear fitting on a graph, and extrapolation (when used in conjunction with a pressure logger sensor and temperature logger sensor - NOT included).
Ordering information: Includes gas study device (55 ml volume), three perforated rubber stoppers, non-perforated rubber stopper, 50 ml syringe, three candles, and activities.
Neulog sensors required (NOT INCLUDED): Pressure logger sensor, temperature logger sensor.
Caution: For educational use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty