Would you like to log in again?
Neulog Light Kit
Supplier: EISCO SCIENTIFIC LLC
LGTKIT
470229-972KT 46 USD InStock
470229-972
Neulog Light Kit
Data Loggers
Hardware for performing NeuLog experiments related to light.
  • Colored light
  • Absorption and reflection of light by different objects
  • Shadows
  • Light intensity and distance
  • Absorption of UV light
The Neulog Light Kit (sensors NOT included) provides all of the necessary tools to study colored light, absorption and reflection of light by different objects, shadows, light intensity and distance, and absorption of UV light by different sunscreens.
Ordering information: Required sensors (not included): Neulog Light sensor, Neulog voltage sensor.
Delivery information: Includes photovoltaic cell, two microscope slides, utility stand, right angle clamp, extension clamp, ruler, sellotape, four colored flashlights (colorless, blue, green, and red), four colored filters (colorless, blue, green, and red), white paper, black.
Caution: For educational use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty