Would you like to log in again?
Neulog Solar Cell Kit
Supplier: EISCO SCIENTIFIC LLC
SLSES
470229-982KT 36.25 USD InStock
470229-982
Neulog Solar Cell Kit
Data Loggers
Hardware to conduct solar power experiments with NeuLog sensors.
  • Kit enables students to study: Photovoltaic cells, light sources, color sensitivity of photovoltaic cells
Uses different light sources (incandescent/fluorescent/sunlight) and color filters to investigate the amount of energy irradiated from a light source, as well as the sensitivity of the solar cell as a function of the color (frequency) of irradiation.
Ordering information: Includes Solar cell, three color filters, flashlight, instructions.
Caution: For educational use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty