Would you like to log in again?
LI-COR® Western Blotting Reagents
Supplier: Li-Cor
926-42212_SE 926-92000_SE 926-95000 926-42213_SE 926-42214_SE 926-91000_SE 926-42211_SE 926-95000_SE 926-98000_SE 926-95010_SE 926-80010_SE 926-80011_SE 926-42210_SE
470230-978EA 446.49 USD InStock
470230-978 470230-988 470230-966 470230-976 470230-986 470230-974 470230-984 470230-972 470230-982 470230-970 470230-980 102971-116 470230-968
LI-COR® Western Blotting Reagents
Blotting Kits
WesternSure® chemiluminescent reagents are optimized for use with film and the C-DiGit® Blot Scanner.
  • WesternSure® chemiluminescent reagents offer the best performance available when compared to other competitive products on the market
WesternSure® PREMIUM is a highly sensitive enhanced substrate for detecting horseradish peroxidase (HRP) on immunoblots.
WesternSure® HRP-conjugated secondary antibodies are compatible with a variety of chemiluminescent substrates and are optimized for use with WesternSure PREMIUM chemiluminescent substrates.
WesternSure® ECL Stripping Buffer allows you to strip and reprobe chemiluminescent Western blots and does not require hazardous shipping, unlike many other stripping buffers.
With the WesternSure® Pen you can annotate visible protein ladders prior to chemiluminescent Western blot detection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty