Would you like to log in again?
Demo Relay Switch
Supplier: EISCO SCIENTIFIC LLC
DER14
470229-994EA 47.95 USD InStock
470229-994
Demo Relay Switch
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Ideal tool for studying and observing the basic concepts of a relay.
  • Relay demonstration device
  • Circuit Board with clear labels
  • Displayed on sturdy base
  • Circuit powering the coil is completely separate from the circuit switched on by the relay
The relay demonstration is an ideal tool for studying and observing the basic concepts of a relay (an electrically operated switch that opens and closes circuits) in a visually and kinesthetically effective manner.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty