Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Assorted Metal Cubes

Supplier: EISCO SCIENTIFIC LLC
PH0106
470230-018EA 10.65 USD InStock
470230-018
Assorted Metal Cubes
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Density Cubes, Set of 6 Metals, 0.4" (10mm) sides.

  • Comprising brass, lead, iron, copper, aluminum, zinc.

Labs Assorted Metal Cubes , Set of six, 10 mm sides, Comprising brass, lead, iron, copper, aluminum, zinc.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty