Would you like to log in again?
Incufridge Pro Environmental Chambers
RS-IF-233PRO RS-IF-253PRO RS-IF-273PRO
470220-200EA 4830 USD InStock
470220-200 470220-198 470220-196
Incufridge Pro Environmental Chambers
Testing Chambers Environmental Test Chambers
The Incufridge Pro line is a series of multi-purpose environmental chamber that can warm, cool, or just hold ambient temperature, depending on your need.
  • Capable of data logging, remote monitoring and alert notifications
  • Two removable metal shelves included
  • Easy to use color touch screen
The Incufridge maintains stable temperatures between 4 to 70 °C, allowing you to heat, cool, or just hold your temperatures at ambient temperature.
All units are Wi-Fi enabled, allowing for critical data logging to be securely stored in the cloud, no expensive software needed. You can program and monitor the device remotely, preventing weekend trips in to check your samples.
The outer door can lock to provide security, and the clear inner door allows you to see your samples while minimizing temperature fluctuations.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty