Would you like to log in again?
Equal Mass, Set 5 cylinders
Supplier: EISCO SCIENTIFIC LLC
PH0112FCB
470230-042EA 27.85 USD InStock
470230-042
Equal Mass, Set 5 cylinders
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
To study the relationship between mass and density.
  • 5 Cylinders, 0.5 inch dia..
  • Aluminum, Copper, Tin, Zinc, Lead
To study the relationship between mass and density. Set of 5 cylinders having same mass and diameter 0.50" varying lengths.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty