Would you like to log in again?
Magnetic Field Demonstrator, 4 Plates
Supplier: EISCO SCIENTIFIC LLC
PH0802MF4
470230-272EA 58.05 USD InStock
470230-272
Magnetic Field Demonstrator, 4 Plates
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
4 Transparent plates to demonstrate magnetic fields.
  • 4 transparent plates to demonstrate magnet fields
  • Each plate is 15.3 cm x 7.6 cm x 0.6 cm - 98 chambers
Comprises a set of four transparent plastic plates 153 x 76 x 6 mm, each plate has a matrix of 98 (14 x 7) small circular chambers, each chamber containing an iron rod and functioning effectively as a plotting compass. The plates may be arranged in various ways, e.g. as a single large two dimensional surface, as faces of a cube etc., and in this way may be used to show configuration of a magnetic field in one or two dimensions. The transparent nature of the plates also makes them suitable for use on an overhead projector. Magnet not included. Picture shows only 2 plates - this actually includes 4 plates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty