Would you like to log in again?
Magnetic Field Demonstration, Large Plate
Supplier: EISCO SCIENTIFIC LLC
PH0802MFD
470230-274EA 46.4 USD InStock
470230-274
Magnetic Field Demonstration, Large Plate
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Excellent tool for viewing magnetic fields.
  • Comprised of a transparent plate with small circular chambers
  • Each chamber contains an iron rod; each functions independently as a plotting compass
  • Transparent- suitable for overhead projectors
Comprising of a transparent plate having a matrix of small circular chambers, each chamber containing an iron rod and functioning independently with as a plotting compass. This plate is an excellent tool to view magnetic fields and due to its transparent nature is suitable for OHPs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty