Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Demonstration Induction Coil

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

PH0840
470230-296EA 51.2 USD InStock
470230-296
Demonstration Induction Coil
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
High quality demonstration induction coil.

  • Both coils fitted with 4 mm sockets for easy use with standard banana plugs
  • Magnetic iron core is 160 mm in length and 15 mm in dia.
  • RoHS compliant and built to last

Primary coil constructed of enameled copper wire, approximate resistance 0.5 Ohms wound on tube with length of 110 x 30 mm in diameter. Secondary coil fits outside primary coil. Coil is constructed of fine enameled copper wire with an approximate resistance of 100 Ohms. Coil is wound on a cylinder with the dimensions 120 mm in length x 82 mm in diameter.

This high quality demonstration induction coil(s) are perfect for showing Faraday's law or incorporating into a bench top research experiment. The set includes 2 nesting coils and cylindrical bar magnet.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty