Would you like to log in again?
Guinea and Feather Apparatus
Supplier: EISCO SCIENTIFIC LLC
PH0160
470230-060EA 51.15 USD InStock
470230-060
Guinea and Feather Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
Compare the falling rate of a feather and a guinea (coin) with air resistance and in a vacuum.
  • Tube is 50 cm long
  • 1mm dia.
  • Includes 40 cm long pressure tubing to connect to vacuum pump (not included)
Glass tube is 50 cm (20") long and 16mm diameter with two rubber bungs, one carrying brass, and 40 cm long pressure rubber tubing to connect to vacuum pump. (1) Glass tube, (1) Piece of Rubber Tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty