Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Make an Ocean Kit

Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
Ward's
1-140
470231-168EA 229 USD InStock
470231-168
Make an Ocean Kit
Educational Materials Biology Educational Materials Ecology Learning Activities
This elementary kit allows students to model four ocean ecology scenarios as they investigate the effect of farm and industrial waste on ocean life.

  • Grade 4-6
  • Class Size 24 students

Brine shrimp populations, water pH and water density are monitored over the course of ecological contamination, water treatment, and recovery. Students also learn about life cycles and how to take representative samples. The kit is ideal for use as a class project or can be completed as a small group. Materials included allow the experiment to be repeated up to five times.

Ordering information: Five 1 gallon aquariums or containers, balance or scale, and a flashlight are required, but not included. Kit contains instructor Manual, MSDS, Life Cycle Chart, Ocean Mat, Medicine Cups, Hydrometer, pH Indicator Paper, Measuring Spoon, Magnifying Glass, Straw, Pipets, Toothpicks, Blank Labels, Slo-Mo Fluid, Brine Shrimp Eggs, Brine Shrimp Food, Farm Runoff Crystals, Industrial Waste Crystals, Chemical Treatment, Sea Salt.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty