Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Plastic Pulley Single, 50 mm Ø

Supplier: EISCO SCIENTIFIC LLC
PH0280A
470230-080EA 5.15 USD InStock
470230-080
Plastic Pulley Single, 50 mm Ø
Educational Materials Physics Educational Materials Mechanics Learning Activities
Economical pulley for simple machine experiments.

  • Single plastic pulley
  • Diameter 50 mm

A high quality, single wheeled plastic pulley, on a steel frame, with hooks at each end and one plastic pulley with a diameter of 50 mm. Inner gap of pulley measures 4 mm, overall length is 8.9 cm. Capable of holding over 30 lbs of weight.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty