Would you like to log in again?
Cell Holders with Terminals on Base
Supplier: EISCO SCIENTIFIC LLC
PH0957C
470230-350EA 27.2 USD InStock
470230-350
Cell Holders with Terminals on Base
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Cell holder for simple circuit.
  • Base is made of folded sheet metal, sockets are RoHS compliant high quality 4 mm banana plug compatible
  • Perfect for constructing simple circuits
The perfect piece of equipment to create power for simple circuits using 4mm banana plugs. Spring loaded connection terminals and secure battery snaps hold (2) D batteries to provide power for any circuit that you build.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty