Would you like to log in again?
Battery Eliminator
Supplier: EISCO SCIENTIFIC LLC
PH0971B
470230-354EA 201.8 USD InStock
470230-354
Battery Eliminator
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Convenient, fixed voltage battery eliminator.
  • Allows teachers to conduct experiments requiring a variety of direct voltages while avoiding the decreased characteristics of dry cells
  • Comprised of a heavy duty transformer wound with copper wire with high lamination to avoid losses
  • Regulated, multiple tapping outputs of 1.5, 3, 4.5, 6, 9, 12 Volts DC
  • Plugs into a standard electrical outlet, 120 V AC, 60 Hz
  • 2 Amps max output
Labs Battery Eliminator, 1.5 – 12 V, max 2 Amps. Tired of hunting for batteries that are the right size or are dead? This convenient, fixed-voltage Battery Eliminator is designed to be safe and reliable source of low voltage, DC power. It is ideal for use in classrooms and substitutes for a wide variety of batteries including AA, AAA, C, D, and 9-volt types. Allows teachers to conduct experiments requiring a variety of direct voltages while avoiding the decreased characteristics of dry cells. Comprised of a heavy duty transformer wound with copper wire with high lamination to avoid losses. Voltage is easily selected with the turn of a dial. Regulated, multiple tapping outputs of 1.5, 3, 4.5, 6, 9, 12 Volts DC. Plugs into a standard electrical outlet, 120 V AC, 60 Hz. 2 amp max output.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty