Would you like to log in again?
Plug Switch One Way
Supplier: EISCO SCIENTIFIC LLC
PH0988A
470230-360EA 9.2 USD InStock
470230-360
Plug Switch One Way
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Plug switch for electric circuits.
  • Perfect for physics experiments or steam punk style setup
  • 4 mm terminals
  • Plastic guard on plug
Unique plug switch that provides an interesting way to complete a circuit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty