Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Density Blocks, Set of 4

Supplier: EISCO SCIENTIFIC LLC
PH0108PCBLF
470230-374KT 53.2 USD InStock
470230-374
Density Blocks, Set of 4
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Density investigation cubes for classroom experiments.

  • Set of 4 - Aluminum, Brass, Copper, Steel
  • 1.25" (3.2cm) Cubes
  • Hooks

This density cube set contains the follow metal cubes: Aluminum, Brass, Copper, Steel. 1.25" (3.2cm) cubes. Hooks installed in each cube.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty