Would you like to log in again?
Economy Electricity Kit
Supplier: EISCO SCIENTIFIC LLC
PH1057
470230-388KT 58.35 USD InStock
470230-388
Economy Electricity Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Ideal for introducing electronics.
  • Everything needed to start learning about basic electricity through hands on assembly of circuits
  • Batteries not included
This kit serves as an ideal starting point for an introduction to electronics. It includes an assortment of circuit components and comes in a plastic storage case.
Ordering information: Kit includes 10 pcs. MES Lamp holders, 10 pcs. MES Bulb, 1 pc. 6 V Buzzer with connecting leads, 1 pc. Electric Motor, 3 - 6 V, 5 pcs. DPDT Slide Switch, 1 pcs. 1.5 V Cell Holder and 10 pcs. Crocodile Leads.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty