Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Decade Resistance Box, 7 decade

Supplier: EISCO SCIENTIFIC LLC
PH1133A7
470230-404EA 263.55 USD InStock
470230-404
Decade Resistance Box, 7 decade
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Ideal for simulating various electrical values.

  • Total Value: 1.111110 mega ohms
  • 7 decades
  • Box length: 19.25in
  • Controller face length: 5in
  • Box heights: narrow side (front): 1.88"/ taller sde (back): 2.75"

The decade resistance box with 7 decades is a sturdy tool that has a total value of 1.111110 Mega Ohms and a tolerance of ±1%, 1 Watt. It is a useful tool for simulating various electrical values.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty