Would you like to log in again?
Decade Resistance Box 4
Supplier: EISCO SCIENTIFIC LLC
PH1133PLN10
470230-406EA 105.55 USD InStock
470230-406
Decade Resistance Box 4
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Standard decade resistance box.
  • Standard decade resistance box
  • Up to a total of 11,110 Ohms
  • Resolution at 1, 10, 100, and 1000 Ohms
  • 0.5 watt resistors
This decade resistance substitution box provides 4 decade ranges as given below and total value 0 to 11110 Ohms with a resolution of 1 Ohm. Each decade is selected using rotary switch. All resistance values are of ±1% tolerance and with 0.5 watt resistors, 0.5555 watts is the theoretical maximum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty