Would you like to log in again?
Stackable Battery Holders
Supplier: EISCO SCIENTIFIC LLC
PH1138AS
470230-410EA 5.5 USD InStock
470230-410
Stackable Battery Holders
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Durable plastic cell holder.
  • Designed to link together in parallel or series to extend capacity or voltage.
  • (1) single holder
  • Made of durable plastic with metal plates for contacts
  • No lead wires
Labs Battery Cell holder - D Size - Stackable Designed to link together in parallel or series to extend capacity or voltage. Made of durable plastic with metal plates for contacts (1) single holder - No lead wires.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty