Would you like to log in again?
Metal Pulley with Clamp
Supplier: EISCO SCIENTIFIC LLC
PH0296A
470230-100EA 19.85 USD InStock
470230-100
Metal Pulley with Clamp
Educational Materials Physics Educational Materials Mechanics Learning Activities
Pulley with metal clamp attached.
  • 50 mm dia. pulley
  • Overall length of frame (pulley bar) 110 mm. Maximum clamp opening 25 mm.
  • Metal Clamp with Plastic pulley
Bench clamp pulley 50 mm diameter pulley with metal ball bearing carried on a two part aluminum frame, designed primarily for clamping to the edge of a bench, table etc. With the pulley overhanging the edge and in a vertical plane. Overall length of frame (pulley bar) 110 mm. Maximum clamp opening 25 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty