Would you like to log in again?
St. Louis Motor Basic
Supplier: EISCO SCIENTIFIC LLC
PH1240N8
470230-434EA 54.2 USD InStock
470230-434
St. Louis Motor Basic
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrates operation of motors and generators.
  • St Louis motor demonstration is useful in demonstrating the operations of motor and generator
  • Consisting of a two-pole armature mounted between magnets
  • Steel supports are mounted on a ridged 4" dia. plastic base
  • Will operate on 3 to 6 V DC
  • Power supply not included
St Louis Motor Demonstration is useful in demonstrating the operations of motor and generator. Consisting of a two-pole armature mounted between magnets. Steel supports are mounted on a ridged 4" Diameter plastic base. Will operate on 3 to 6 V DC. Power supply not included. This open design allows students to see each part of the motor operate, which shows basic function.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty