Would you like to log in again?
Metal Pulley with Clamp
Supplier: EISCO SCIENTIFIC LLC
PH0301A
470230-110EA 30.95 USD InStock
470230-110
Metal Pulley with Clamp
Educational Materials Physics Educational Materials Mechanics Learning Activities
Plastic pulley with metal clamp, includes knob for tightening.
  • Clamp made from high quality metal
  • Painted red
  • Pulley made of high quality plastic
  • Grey hand knob for tightening clamp
Clamp has a 45 mm plastic pulley with bracket for bench mounting up to 25mm thickness. Can be used with a force table as well. Wood in picture not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty