Would you like to log in again?
Pendulum Clamp Aluminum
Supplier: EISCO SCIENTIFIC LLC
PH0308
470230-114EA 30.95 USD InStock
470230-114
Pendulum Clamp Aluminum
Educational Materials Physics Educational Materials Mechanics Learning Activities
Aluminum pendulum clamp with screw clamps for attaching to rods.
  • For use in demonstrations or experiments involving pendulums
  • Can attach up to three pendulums to three screw clamps
  • Clamp attaches to support rods up to 16 mm in dia.
This sturdy pendulum clamp is constructed of an aluminum support bar with three screw clamps, attached to a die cast head clamp for use on support rods to hang weights and pendulums. Can attach to support rods up to 16mm in diameter. Three screw clamps can securely and individually be used to attach weights with strings. This pendulum clamp makes it easy to simultaneously demonstrate or test the properties of simple harmonic motion in physics labs or classrooms.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty