Would you like to log in again?
Bell Demonstration Model
Supplier: EISCO SCIENTIFIC LLC
PH1272
470230-438EA 45.75 USD InStock
470230-438
Bell Demonstration Model
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Displays basic principles of electromagnetism.
  • This labs electromagnet bell shows the basic principles of electromagnetism.
  • To activate the bell, there is a push button to complete the circuit.
  • All components are visible along with wiring so students can understand the operation of this model.
  • All pieces are made of rigid materials which are mounted on a metal plate which is attached to a 8" x 4" wooden base
  • Power supply is required, but sold separately
This Labs electromagnet bell shows the basic principles of electromagnetism. The model is equipped with two 4 mm banana plugs outlets for a external power supply to operate this demonstration. To activate the bell, there is a push button to complete the circuit. All components are visible along with wiring so students can understand the operation of this model. All pieces are made of rigid materials which are mounted on a metal plate which is attached to a 8" x 4" wooden base. This stands approx. 11" Tall.
Ordering information: Power supply is not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty