Would you like to log in again?
Analogue to Digital Convertor
Supplier: EISCO SCIENTIFIC LLC
PH1366
470230-448EA 121.35 USD InStock
470230-448
Analogue to Digital Convertor
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrates basis of digital communications.
  • Converts analog signal to a binary digital output
  • Great for demonstrating the basis of digital communications
  • Companion apparatus to the labs digital to analog converter
An excellent system for demonstrating the basis of digital communications. An analogue input in the range 0 to 7 V is converted to a binary digital output in 1 V steps (i.e. 1 V is represented by 001, 2 V by 010, etc.). A switch converts the unit to 0 to 2 V sensitivity so that low frequency sine waves from a signal generator can be digitized. The binary output is available at 4 mm sockets for use with the companion Labs Digital to Analog Converter (PH1367). A 12 V DC input is required from a separate power supply.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty