Would you like to log in again?
Inclined Plane Steel Basic
Supplier: EISCO SCIENTIFIC LLC
PH0338
470230-126EA 78 USD InStock
470230-126
Inclined Plane Steel Basic
Educational Materials Physics Educational Materials Mechanics Learning Activities
Steel inclined plane with accessories.
  • Designed specially for students
  • Made entirely of heavy gauge steel
  • Plane can be clamped at any angle from 0 to 45
  • Board dimensions 600 x 100mm
  • Pan and roller included
  • Complete with instructions
Designed specially for students. Made entirely of heavy gauge steel, the base and plane are connected by a hinge, allowing the plane to be clamped at any angle from 0 to 45. Vertical displacement, angular displacement and distance from the hinge can be measured with built in scales. The pulley can be adjusted to ensure that the cord runs parallel to the plane. Board dimensions 600 x 100 mm. Pan and roller included. Complete with instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty