Would you like to log in again?
Force Table, Economy
Supplier: EISCO SCIENTIFIC LLC
PH0347FT
470230-142EA 180.6 USD InStock
470230-142
Force Table, Economy
Educational Materials Physics Educational Materials Mechanics Learning Activities
Force table is inexpensive and light in weight.
  • Three-legged
  • Top is made from durable PVC
Laminated top allows use of erasable markers. The legs screw in the table quite easily and have built-in levelling screws for levelling adjustments.
Ordering information: Weights not included.
Delivery information: The table is supplied with 4 special ball bearing pulleys and 4 weight hangers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty