Would you like to log in again?
Maxwell Apparatus with Stand
Supplier: EISCO SCIENTIFIC LLC
PH0349
470230-144EA 69.3 USD InStock
470230-144
Maxwell Apparatus with Stand
Educational Materials Physics Educational Materials Mechanics Learning Activities
Maxwell apparatus with flywheel to demonstrate momentum, friction, and oscillation.
  • Constructed of heavy wooden base and solid metal fly wheel
  • Flywheel cord can be easily adjust from the top to fine tune results
  • Wind cords around flywheel axle and release. The wheel will unwind then recoil due to momentum
  • Flywheel can adjusted off center to examine effects.
  • Oscillation will run for quite some time due to the adjustability of the apparatus
  • Eventually loss of momentum and friction will cause the flywheel to come to rest
Maxwell Wheel Apparatus With Easy Adjustment With 22" Height And 3.5" Diameter Fly Wheel. Constructed of heavy wooden base and solid metal fly wheel. Flywheel cord can be easily adjust from the top to fine tune results. Wind cords around flywheel axle and release. The wheel will unwind then recoil due to momentum. Flywheel can adjusted off center to examine effects. Oscillation will run for quite some time due to the adjustability of the apparatus. Eventually loss of momentum and friction will cause the flywheel to come to rest
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty