Would you like to log in again?
Dynamics Cart, Set 3 Wheeled
Supplier: EISCO SCIENTIFIC LLC
PH0350A
470230-150PR 148.05 USD InStock
470230-150
Dynamics Cart, Set 3 Wheeled
Educational Materials Physics Educational Materials Mechanics Learning Activities
Low friction cart for kinematics.
  • Carts have with low-friction ball-bearing wheels
This pair of cart is constructed from solid wood and aluminum. Fitted with high quality frictionless nylon ball bearing wheels allows for more accurate dynamic measurements. One of the carts is equipped with an exploder piston that is activated by a trigger on the top of the cart.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty