Would you like to log in again?
Pipe Thread Nipples, Western Enterprises
Supplier: Western Enterprises
312-BN-4-50HP 312-BN-4-35HP 312-BN-4-15HP 312-BN-4-30HP 312-BN-4-20HP
300066-668EA 28.91 USD InStock
300066-668 300066-664 300066-666 300007-946 300066-662
Pipe Thread Nipples, Western Enterprises
Welding Supplies
Pipe thread nipples are used to obtain a threaded connection on the end of standard scheduled pipe sizes rather than O.D
tube sizes, so that NPT pipe threaded fittings may be attached. These nipples are held in place with a standard compression coupling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty