Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bar Breaking Apparatus

Supplier: EISCO SCIENTIFIC LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

PH0380A
470230-162EA 81.5 USD InStock
470230-162
Bar Breaking Apparatus
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
This apparatus is useful for studying forces during thermal contraction and expansion.

  • 13" long bar
  • Contains iron base, bar with tension nut, and breaking bars

The bar breaking apparatus provides a very sturdy and user-friendly method by which to introduce students to the forces that can occur during thermal contraction and thermal expansion. Students can experience and study the forces of thermal contraction.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty