Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hand Held Centripetal Force Kit

Supplier: EISCO SCIENTIFIC LLC
PH1120152
470230-402KT 11.3 USD InStock
470230-402
Hand Held Centripetal Force Kit
Educational Materials Physics Educational Materials Mechanics Learning Activities
Ideal for studying rotational dynamics.

  • Kit Contains: (1) Hollow plastic grip handle, (1) String, (20) Washers, (2) Paper Clips, (1) Rubber stopper
  • Student puts string through handle, spins rubber stopper around and balances that with the masses at the bottom of the string
  • Activity allows students to study how mass and radius affect centripetal acceleration and rotational velocity
  • Easy fund demonstration of centripetal force

This elegantly simple kit provides the necessary tools to discover properties of rotational dynamics. A rubber stopper swings around on the end of a string counter balanced by weights, allowing students to see how mass and radius affect centripetal acceleration and rotational velocity.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty