Would you like to log in again?
Static Electricity Accessories
Supplier: EISCO SCIENTIFIC LLC
PH0918C
470230-322KT 47.6 USD InStock
470230-322
Static Electricity Accessories
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Accessories for electrostatic generators.
Put your electrostatic generator (van de Graaff or Wimshurst) to use to do more than create static charge. Physically demonstrate numerous effects with this kit.
Ordering information: Kit includes (1) Dancing Ball Cylinder, (1) Plume of Hair with connector, (1) Plastic Comb, (1) Point Source Needle, (1) Neon Light Indicator, (1) Spinning Spiked Arm Wheel and (1) Pith ball on String and Cup.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty