Would you like to log in again?
Accessories for Van de Graaff
Supplier: EISCO SCIENTIFIC LLC
PH0920C
470230-328KT 45 USD InStock
470230-328
Accessories for Van de Graaff
Educational Materials Physics Educational Materials
Accessories for Van de Graaff.
  • Items with in Set, Pillar with suspended metallized sphere, Faraday’s Pail
  • Perspex cylinder with metal caps and 4 metallized spheres, Head of hair, Point discharger, Neon bulb pointer, Electric whirl, Comb, Banana Plug ( 34" ), Spare O ring Belt
  • Recommended use with PH0918A, PH0920A, or EDUVDG
  • Put your electrostatic generator (van de Graaff or Wimshurst) to use to do more than create static charge
  • Physically demonstrate numerous effects with this kit
  • No instructions included
The accessories may be fixed in 4 mm. sockets in top of sphere. Items with in Set, Pillar with suspended metallized sphere, Faraday's Pail. Perspex cylinder with metal caps and 4 metallized spheres, Head of hair, Point discharger, Neon bulb pointer, Electric whirl, Comb, Banana Plug ( 34" ), Spare O ring Belt.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty