Would you like to log in again?
Telescope Model
Supplier: EISCO SCIENTIFIC LLC
PH0605A
470230-206KT 39 USD InStock
470230-206
Telescope Model
Educational Materials Physics Educational Materials Optics Learning Activities
Functional model of telescope.
  • Functional telescope model
  • Rugged PVC construction
  • Co-axially mounted tubes extend to 21.5" length
  • Perfect demonstration tool for telescope function
  • 5X magnification
This is a functional and durable PVC telescope model that is great for demonstrations and classroom use as well as a fun tool for home use. It is compact, lightweight and easy to store. The model provides 5× magnification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty